Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-1236-01

ExcelGel XL SDS 12-14

Cytiva 17-1236-01, pkg of 3 pieces

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

form

solid

feature

wetted part: no

packaging

pkg of 3 pieces

manufacturer/tradename

Cytiva 17-1236-01

color

Colorless

Application

The new Multiphor II Buffer Strip Positioner facilitates placement of ExcelGel Buffer Strips and holds the strips securely in place during the run.

Precast polyacrylamide gels make a wide range of electrophoretic techniques readily available with minimum preparation. The manufacturing quality and reproducibility of these gels translate into highly reliable and reproducible results with minimal training.

ExcelGel SDS and Ampholine® PAGplate products are supplied fully hydrated and ready to use. CleanGel, Immobiline® DryPlate, and DryStrip products are supplied dry and must be rehydrated in the appropriate buffer before use. Prepared swelling solutions are available for the most common procedures. All precast gel recipes are proprietary. However, buffer formulations for a variety of other applications are included in the instructions.

Features and Benefits

  • Perform second-dimension separations using convenient precast gels and 7-, 11-, 18-, or 24-cm IPG strips.
  • Choose from ExcelGel precast SDS-polyacrylamide gels available as 12.5% homogeneous and 12%–14% gradient gels.
  • Prepare multiple 2-D maps on a single gel using three 7-cm or two 11-cm IPG strips side-by-side.
  • Configure Multiphor II with MultiTemp III Thermostatic Circulator and the programmable EPS 1001 power supply for reproducible running conditions.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C ()

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Ampholine is a registered trademark of Cytiva
Immobiline is a registered trademark of Cytiva

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

This page describes how to apply equilibrated Immobiline® DryStrip gels on the ExcelGel SDS gels.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service