Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-0885-02

Gammabind G Sepharose

Cytiva 17-0885-02, pack of 25 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13111000
NACRES:
NA.56

ligand

recombinant GammaBind G type 2 lacking albumin-binding region

packaging

pack of 25 mL

manufacturer/tradename

Cytiva 17-0885-02

storage condition

(20% Ehtanol, PBS pH 7)

matrix

4% agarose

particle size

45-165 μm

average diameter

90 μm

cleaning in place

2-9

working range

3-9

capacity

>17 mg binding capacity(human IgG/ml)

storage temp.

2-8°C

General description

Recombinant GammaBind is an affinity medium for laboratory scale antibody purification. Binds to all IgG subclasses from human, mouse, and rat; binds total IgG from guinea pig, rabbit, goat, cow, sheep, and horse.

GammaBind G Sepharose is GammaBind G, Type 2, covalently immobilized to Sepharose 4B by the cyanogen bromide method. GammaBind G, Type 2, a recombinant form of streptococcal Protein G, binds to the Fc region of IgG from a variety of mammalian species. GammaBind G Sepharose may be used to analyze and purify classes, subclasses and fragments of immunoglobulins from any biological fluid or cell culture medium.

GammaBind G Sepharose may be used to analyze and purify classes, subclasses and fragments of immunoglobulins from any biological fluid or cell culture medium. Since only the Fc region is involved in binding, the Fab region is still available for binding antigen. Hence, GammaBind G Sepharose is extremely useful for isolation of immune complexes. GammaBind G will not cross-react with other serum proteins such as IgM, IgE, IgA or transferrin.
pH below 3 is sometimes required to elute strongly bound IgG species. However, protein ligands may hydrolyze at very low pH.

Application

The potential applications of GammaBind G Sepharose include practically all of the current and projected applications of Protein A Sepharose. However, GammaBind G and Protein A have different IgG binding specificities, depending on the origin of the IgG. Compared to Protein A, GammaBind G binds more strongly to mouse and rat monoclonal IgG and to IgG from goat, sheep, horse, cow, human, rabbit and other mammalian species.

Features and Benefits

  • Recombinant GammaBind G type 2 coupled to Sepharose 4B engineered for antibody purification at laboratory scale.
  • Binds to all IgG subclasses from human, mouse, and rat.
  • Binds total IgG from guinea pig, rabbit, goat, cow sheep, and horse.
  • Will not cross-react with other serum proteins such as IgM, IgE, IgA or transferrin.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

pictograms

Flame

signalword

Warning

hcodes

Storage Class

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service