Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-0872-50

Protein A

Cytiva, Freeze Dried

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

feature

Freeze Dried

manufacturer/tradename

17-0872-50
Cytiva

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

"" - ""

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service