Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-0847-02

CSTFA

Cytiva 17-0847-02

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.52

packaging

pkg of 100 mL

manufacturer/tradename

Cytiva 17-0847-02

storage temp.

2-8°C

General description

illustra CsTFA is a strong chaotrope that not only inhibits ribonucleases, but also promotes the hydration and solubilization of nucleic acids and proteins. Due to its high solubility, solutions can be made with densities of up to 2.6 g/mL.

Application

illustra CsTFA (solution*) is a strong chaotrope that not only inhibits ribonucleases, but also promotes the hydration and solubilization of nucleic acids and Proteins. Due to its high solubility, solutions can be made with densities of up to 2.6 g/mL. This feature, combined with the fact that nucleic acids have relatively Low buoyant densities in CsTFA (as compared with CsCl), means that RNA can be banded isopycnically in CsTFA gradients. Conditions can also be chosen in which RNA pellets at the bottom of the gradient while DNA is banded, thus allowing very effective purification of RNA from DNA.

Features and Benefits

  • Effective inhibitor of ribonucleases
  • Can separate RNA and DNA.
  • Soluble in ethanol so nucleic acids can be recovered from the gradient by ethanol precipitation.
  • Strong chaotrope.
  • Promotes the hydration and solubilization of nucleic acids and Proteins.
  • Designed to ensure high reproducibility, yield, and purity with every experiment.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

illustra is a trademark of Cytiva

pictograms

Exclamation mark

signalword

Warning

Storage Class

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service