Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-0700-01

2′,5′ ADP-Sepharose 4B

Cytiva 17-0700-01, pack of 5 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

packaging

pack of 5 g

manufacturer/tradename

Cytiva 17-0700-01

matrix

4% agarose

particle size

45-165 μm

average diameter

90 μm

cleaning in place

4-10

working range

4-10

shipped in

wet ice

storage temp.

2-8°C

General description

2′5′ ADP Sepharose 4B media with immobilized NADP structural analog is primarily used for purification of enzymes requiring NADP as a cofactor.

Application

2′5′ ADP Sepharose 4B interacts strongly with NADP+-dependent dehydrogenases. Selective elution with gradients of NAD+ or NADP+ has allowed the resolution of complex mixtures of dehydrogenase isoenzymes using 2′5′ ADP Sepharose 4B.

Features and Benefits

  • 2′5′ ADP is an NADP structural analog immobilized on Sepharose 4B. It binds and immobilizes enzymes requiring this cofactor

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Storage Class

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows how to purify NADP+-dependent dehydrogenases and other enzymes with affinity for NADP+ by affinity chromatography using 2’5’ ADP Sepharose 4B and Red Sepharose CL-6B from Cytiva.

This page shows how to convert between flow velocity and volumetric flow rate in affinity chromatography of antibodies.

This page shows how to solve practical problems that may occur when running an affinity chromatography column.

Protocols

This page shows how to perform column packing and preparation for affinity chromatography when using Tricorn™ or XK columns available from Cytiva.

This page shows how to prepare samples for purification with affinity chromatography.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service