Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-0516-01

PhastGel® Buffer Strips

3% Agarose IEF, SDS, pack of 20 strips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105300
NACRES:
NB.22

form

solid

packaging

pack of 20 strips

manufacturer/tradename

Cytiva 17051601

size

41 mm × 10 mm × 6 mm

color

Colorless

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

  • Select from homogeneous and gradient polyacrylamide gels.
  • Separate proteins and peptides on denaturing, native, and isoelectric focusing gels.
  • Save time and improve reproducibility with precast gels, buffer strips, and staining kits.
  • Achieve detection sensitivity comparable to that of radioisotopes with ready-to-use silver staining kits.

PhastGel Buffer Strips are available in three types – PhastGel DNA, PhastGel SDS, and PhastGel Native. All are supplied as ready-to-use agarose blocks, making buffer preparation unnecessary. PhastGel DNA Buffer Strips are specifically tested for DNA separations. When used with PhastGel DNA Silver Staining Kit, they improve the sensitivity of detection.

Physical form

0.20 M tricine, 0.20 M tris, 0.55% SDS, pH 8.1

Legal Information

PhastGel is a registered trademark of Cytiva

Storage Class

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service