Key Documents
GE11-0004-21
Ion Exchange Chromatography And Chromatofocusing Handbook
Cytiva 11-0004-21
About This Item
UNSPSC Code:
12352200
packaging
pkg of 1 ea
manufacturer/tradename
Cytiva 11-0004-21
General description
This Handbook contains the latest information on the theoretical and practical aspects of ion exchange and chromatofocusing techniques, the prepacked columns and media available, and how to select them. The Handbook provides many application examples, practical information together with hints and tips on improving ion exchange chromatography and chromatofocusing protocols.
Storage Class
13 - Non Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)