GE10600127
Amersham™ Protran® supported western blotting membrane for use with Minifold I blot-spot system
nitrocellulose membrane, pore size 0.45 μm, L × W 102 mm × 133 mm, sheets, pack of 10 ea
Synonym(s):
blotting and detection, blotting membrane
All Photos(1)
About This Item
UNSPSC Code:
41100000
material
nitrocellulose membrane
sheets
packaging
pack of 10 ea
manufacturer/tradename
Cytiva 10600127
Cytiva Article No. 29-0912-77 (US reference)
L × W
102 mm × 133 mm
pore size
0.45 μm pore size
General description
Nitrocellulose membrane suitable for proteins of a wide range of molecular weights. Low background and excellent signal-to-noise ratios, compatible with a variety of labeling and detection systems, high binding capacicity and proprietary nitrocellulose formula gives a long shelf life of bound proteins
Amersham Protran offers excellent sensitivity, resolution, and low background, especially when used with Amersham ECL, Amersham ECL Prime, or Amersham ECL Select Western Blotting Systems.
Amersham Protran offers excellent sensitivity, resolution, and low background, especially when used with Amersham ECL, Amersham ECL Prime, or Amersham ECL Select Western Blotting Systems.
Legal Information
Amersham is a trademark of Cytiva
Protran is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)