Key Documents
GE10600061
Amersham Hybond® SEQ Western blotting membranes, PVDF
pore size 0.2 μm, sheet W × L 200 mm × 200 mm, pkg of 10 ea
About This Item
UNSPSC Code:
41100000
packaging
pkg of 10 ea
manufacturer/tradename
Cytiva 10600061
sheet W × L
200 mm × 200 mm
pore size
0.2 μm pore size
General description
PVDF protein membrane, specifically designed for protein sequencing applications and solid phase amino acid analysis.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Legal Information
Amersham is a trademark of Cytiva
Hybond is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)