GBL910064
Grace Bio-Labs silicone gasket for ProPlate® microarray system
fits 64 wells, size 3.5 mm × 3.5 mm
Synonym(s):
microarray chambers for slides, silicone slide gasket
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
sterility
non-sterile
manufacturer/tradename
Grace Bio-Labs 910064
size
3.5 mm × 3.5 mm
64 wells
color
none
Looking for similar products? Visit Product Comparison Guide
General description
Replacement Silicone Gaskets for Grace BioLabs Proplate slide modules.
While the silicone gaskets affixed to wells may be re-used multiple times, their lifespan may be shortened by washing conditions, repeated autoclaving and prolonged exposure to harsher types of chemicals. Therefore, replacement gaskets are available and may be easily positioned and applied to the chambers after simply peeling the old gasket off and throwing it away.
While the silicone gaskets affixed to wells may be re-used multiple times, their lifespan may be shortened by washing conditions, repeated autoclaving and prolonged exposure to harsher types of chemicals. Therefore, replacement gaskets are available and may be easily positioned and applied to the chambers after simply peeling the old gasket off and throwing it away.
Application
- Protein and peptides microarrays
- cDNA and oligonucleotides microarrays
- Proteomics
- Protein expression analysis
- Antibody profiling
Legal Information
ProPlate is a registered trademark of Grace Bio-Labs, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service