GBL664103
Grace Bio-Labs Press-To-Seal silicone isolator, No PSA
wells, 1 (Rounded rectangle, 19mm x 32mm x 0.8mm depth)
Synonym(s):
Nonadhesive reagent isolators, autoclavable slide chambers, removable hydrophobic barrier
Select a Size
$150.00
Available to ship onApril 29, 2025Details
Select a Size
About This Item
$150.00
Available to ship onApril 29, 2025Details
Recommended Products
material
red silicone
sterility
non-sterile
feature
wells 1 (Rounded rectangle, 19mm x 32mm x 0.8mm depth)
packaging
pack of 25 ea
manufacturer/tradename
Grace Bio-Labs 664103
external L × W
44 mm × 25.5 mm
Looking for similar products? Visit Product Comparison Guide
General description
Silicone Isolators remain sealed to smooth surfaces during washing steps. Where additional sealing is required, SecureSeal adhesive on one or both surfaces is recommended. Each seal design is also available in CultureWell "water-clear" silicone, which is highly recommended for sensitive cell culture applications. Closed chambers may be formed using flexible RNase free, HybriSlip covers. Recommended use temperature is -62°C to 218°C.
Application
Live-cell imaging
Microscoscopy
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service