Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GBL204996

Grace Bio-Labs FlexPlate MP microtiter plate superstructure

size 96 wells, L × W 7.25 mm × 7.25 mm, clear silicone, adhesive one side

Synonym(s):

incubation chamber cover, silicone microtiter plate cover

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
material:
clear silicone
colorless wells
size:
96 wells

material

clear silicone
colorless wells

sterility

non-sterile

feature

lid: no

packaging

pack of 1 ea

manufacturer/tradename

Grace Bio-Labs 204996

L × W

7.25 mm × 7.25 mm

size

96 wells

color

clear

Looking for similar products? Visit Product Comparison Guide

General description

N/A

Legal Information

FlexPlate MP is a trademark of Grace Bio-Labs, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service