Key Documents
GBL104418
Grace Bio-Labs MultiSlip coverglass inserts, with chambered coverglass
L × W 18 mm × 18 mm, sterile, wells, 8
Synonym(s):
chamber slides, chambered coverglass, cover slides, incubation chambers
About This Item
UNSPSC Code:
41122600
NACRES:
NB.22
material
colorless
silicone (on glass)
sterility
sterile
feature
wells 8
manufacturer/tradename
Grace Bio-Labs 104418
L × W
18 mm × 18 mm
well area
3.24 cm2
wells
8
General description
Grace Bio-Labs MultiSlip insert with 8 (18 mm x 18 mm) or 15 (12 mm x 12 mm) number 1.5 German coverglass per insert are sterile and ready to use in conventional 86 mm x 128 mm culture plates for cell culture applications. Staining and washing procedures may be performed with MultiSlip inserts in the plate, or silicone backed coverglass may be removed individually and affixed to glass microscope slides. Alternately, inserts may be easily removed for batch processing in glass staining dishes.
Note: MultiSlip coverglass inserts are ideally suited for the culture of cells where pretreatment of glass surfaces with a biological coating is required.
Legal Information
Multislip is a trademark of Grace Bio-Labs, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)