Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G9790

Sigma-Aldrich

Anti-Glutamate Receptor 2/3, Metabotropic (mGluR2/3) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, feline, mouse

technique(s)

immunohistochemistry (frozen sections): 1:500
immunoprecipitation (IP): 1:100
indirect immunofluorescence: 1:500
western blot: 1:1,000

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

Gene Information

mouse ... Grm2(108068)
rat ... Grm2(24415)

General description

The metabotropic glutamate receptors consist of eight subtypes (mGluR1-8) divided into three groups (I-III). Glutamate is the main excitatory neurotransmitter in the brain. It acts on ligand-gated receptor channels, termed NMDA, AMPA and kainate receptors, involved in the fast excitatory synaptic transmission. Glutamate also regulates ion channels and enzymes producing second messengers via specific receptors coupled to G-proteins, called metabotropic glutamate receptors.

Specificity

This antibody recognizes the C-terminal sequence shared by metabotropic glutamate receptors 2 and 3.

Immunogen

synthetic peptide corresponding to the C-terminal region (13 amino acids) of mGluR2/3.

Application

Anti-Glutamate Receptor 2/3, Metabotropic (mGluR2/3) antibody produced in rabbit is suitable for immunohistochemistry and indirect immunofluorescence at a working dilution of 1:500. It is suitable for immunoprecipitation at 1:100 and for immunoblotting at 1:1000 working dilution.

Biochem/physiol Actions

GRM2 and GRM3 belong to group II receptors and are involved in the inhibition of the cyclic AMP cascade. The role of the metabotropic receptors in pain and pathology is being studied for therapeutic potential. Glutamate receptors are key regulators of excitatory amino acid neurotransmission in the striatum.

Physical form

Solution in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100μg/mL BSA, and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Makoff et al.
Brain research. Molecular brain research, 40(1), 55-63 (1996-08-01)
A human brain cDNA library was screened using amplified human metabotropic glutamate receptor (mGluR) cDNA sequences as probes. The resulting clones included one containing the complete coding sequence of mGluR3. This sequence has 90% DNA sequence identity with rat mGluR3
Carla Letizia Busceti et al.
eNeuro, 11(8) (2024-07-06)
Cognitive dysfunction is associated with methamphetamine use disorder (MUD). Here, we used genetic and pharmacological approaches to examine the involvement of either Group 2 metabotropic glutamate (mGlu2) or mGlu3 receptors in memory deficit induced by methamphetamine in mice. Methamphetamine treatment
P J Flor et al.
The European journal of neuroscience, 7(4), 622-629 (1995-04-01)
A cDNA encoding the human metabotropic glutamate receptor type 2 (hmGluR2) was isolated from human brain cDNA libraries by cross-hybridization with rat mGluR2 probes. The deduced amino acid sequence of the human mGluR2 receptor consists of 872 residues and shows
W Spooren et al.
Behavioural pharmacology, 14(4), 257-277 (2003-07-03)
Following the molecular cloning in the early 1990s of the metabotropic glutamate receptors (mGlu1-8), research that focused on the physiology, pharmacology and function of these receptors revealed their potential role in CNS disorders. Numerous psychiatric and neurological dis-orders are indeed
J P Pin et al.
Neuropharmacology, 34(1), 1-26 (1995-01-01)
Glutamate is the main excitatory neurotransmitter in the brain. For many years it has been considered to act only on ligand-gated receptor channels--termed NMDA, AMPA and kainate receptors--involved in the fast excitatory synaptic transmission. Recently, glutamate has been shown to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service