Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G9759

Sigma-Aldrich

Anti-Goat Serum antibody produced in rabbit

fractionated antiserum, lyophilized powder

Synonym(s):

Anti-Serum, goat

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

fractionated antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

goat

packaging

vial of 2 mL lyophilized antiserum

technique(s)

immunoelectrophoresis: suitable

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Physical form

Lyophilized from 0.01 M phosphate buffered saline, pH 7.2

Reconstitution

Reconstitute with 2 ml deionized water.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maciej J Smialek et al.
Cells, 9(4) (2020-04-23)
Mammalian Pumilio (PUM) proteins are sequence-specific, RNA-binding proteins (RBPs) with wide-ranging roles. They are involved in germ cell development, which has functional implications in development and fertility. Although human PUM1 and PUM2 are closely related to each other and recognize

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service