Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G9670

Sigma-Aldrich

Monoclonal Anti-O-Glycosylated Neurofilament-M antibody produced in mouse

~2 mg/mL, clone NL6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-O-glycosylated NF-M

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

NL6, monoclonal

form

buffered aqueous solution

mol wt

antigen ~160 kDa

species reactivity

human, rat

should not react with

bovine, mouse

packaging

antibody small pack of 25 μL

concentration

~2 mg/mL

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: 0.5-1 μg/mL using human NT2 cell extracts

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NEFM(4741)
rat ... Nefm(24588)

General description

Intermediate filaments (IFs) have a diameter of 8-10nm and differ from microtubules and microfilaments significantly. Neurofilaments (NFs) are a type of IFs found mainly in cells or tissues of neuronal origin. NFs are composed of three major proteins, NF-L, NF-M and NF-H, having a molar mass of 68kDa, 160kDa, and 200kDa, respectively.
Monoclonal Anti-O-Glycosylated Neurofilament-M (mouse IgG2a isotype) is derived from the hybridoma NL6 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a protein fraction enriched for components of the human neuronal cytoskeleton. Neurofilament medium (NEFM) gene encodes neurofilament medium polypeptide which constitutes to cytoskeleton of mature neurons. The gene in human chromosome is localized on 8p21.

Immunogen

protein fraction enriched for components of the human neuronal cytoskeleton from cell lines NT2-N and SK-N-BE(2). The epitope is localized in the carboxy-terminal tail domain of NF-M.

Application

Monoclonal Anti-O-Glycosylated Neurofilament-M antibody produced in mouse has been used in:
  • immunohistochemical methods
  • immunoblotting
  • immunocytochemistry

Biochem/physiol Actions

Neurofilament medium (NEFM) is essential for radial growth of axon which includes the diameter of the axon as well as the myelination. The C-terminus of NFEM gene has 7 repeats of KSP (lysine-serine-proline) domain and phosphorylation of which is crucial for regulating the radial growth of axon.
The NF proteins are synthesized in the neuronal perikarya before assemby to form the filaments. These filaments are then transported within the axons towards the synaptic terminals. The function of these NF polypeptides is regulated by phosphorylation, which influences their interaction with cytoplasmic organelles. The head region of NF-L and NF-M contain sites modified by O-linked GlcNAc. Down-regulation of NF-M O-GlcNAcylation and increased phosphorylation of NF-M is observed in Alzheimer disease. This antibody recognizes specifically an O-glycosylated epitope in the projection domain of NF-M. In a rat model for amyotrophic lateral sclerosis (ALS), this specific modification is strongly decreased compared with wild-type animals.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A García-Sevilla et al.
Neuroreport, 8(7), 1561-1565 (1997-05-06)
NEUROFILAMENT (NF) proteins, the major components of the neuronal cytoskeleton, have been shown to represent previously unknown targets for the chronic effects of morphine in rats. This study was designed to evaluate the abundance of immunoreactive NF-L (68 kDa) proteins
J Q Trojanowski et al.
Annals of medicine, 21(2), 113-116 (1989-01-01)
Here we review our recent "epitope analyses" of a few of the fibrous intraneuronal inclusions that are distinctive hallmarks of human neurodenerative conditions using a large library of monoclonal antibodies (MAbs) raised to normal neuronal cytoskeletal proteins. Analyses of the
Phosphorylation of neuronal cytoskeletal proteins in Alzheimer's disease and Lewy body dementias.
J Q Trojanowski et al.
Annals of the New York Academy of Sciences, 747, 92-109 (1994-12-15)
Yanqiu Deng et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 22(1), 138-145 (2007-08-10)
The medium subunit of neurofilament (NF-M) is extensively modified by phosphate and O-linked beta-N-acetylglucosamine (O-GlcNAc). Phosphorylation of NF-M plays a critical role in regulating its translocation, filament formation, and function. However, the regulation of NF-M phosphorylation and the role of
NF-M is an essential target for the myelin-directed ?outside-in? signaling cascade that mediates radial axonal growth
Garcia ML, et al.
The Journal of Cell Biology, 163(5), 1011-1020 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service