Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

G9267

Sigma-Aldrich

Glutathionesulfonic acid

Synonym(s):

Glutathione sulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17N3O9S
CAS Number:
Molecular Weight:
355.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

assay

≥98% (TLC)

form

powder

technique(s)

ligand binding assay: suitable

color

white

storage temp.

2-8°C

SMILES string

NC(CCC(=O)NC(CS(O)(=O)=O)C(=O)NCC(O)=O)C(O)=O

InChI

1S/C10H17N3O9S/c11-5(10(18)19)1-2-7(14)13-6(4-23(20,21)22)9(17)12-3-8(15)16/h5-6H,1-4,11H2,(H,12,17)(H,13,14)(H,15,16)(H,18,19)(H,20,21,22)

InChI key

QGWRMTHFAZVWAM-UHFFFAOYSA-N

Amino Acid Sequence

Glu-Cys-Gly

Biochem/physiol Actions

Glutathionesulfonic acid (GSO3), a metabolite of glutathione, is used to increase the tissue-to-plasma concentration ratio of thiopental sodium. Glutathione sulfonate is also used as a modulator of dopamine release and to study glutathione binding sites on proteins such as glutathione transferase.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Réka Janáky et al.
Neurochemical research, 32(8), 1357-1364 (2007-04-03)
Glutathione (gamma-glutamylcysteinylglycine, GSH and oxidized glutathione, GSSG), may function as a neuromodulator at the glutamate receptors and as a neurotransmitter at its own receptors. We studied now the effects of GSH, GSSG, glutathione derivatives and thiol redox agents on the
Yuhsuke Ohkawa et al.
Biological & pharmaceutical bulletin, 25(6), 761-765 (2002-06-26)
The effects of glutathione (GSH) and glutathionesulfonic acid sodium salt [N-(N-gamma-L-glutamyl-L-beta-sulfoalanyl)glycine sodium salt, GSO3Na], which is a minor metabolite of GSH, on the pharmacokinetics of thiopental sodium were investigated in rats. The concomitant use of GSO3Na with thiopental sodium significantly
David Balchin et al.
Biophysical chemistry, 156(2-3), 153-158 (2011-05-03)
A C-terminal helix (α9) adjacent to the active site on each subunit is a structural feature unique to the alpha isoform of glutathione transferases which contributes to the catalytic and ligandin functions of the enzyme. The ionisation state of Tyr-9

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service