Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G8895

Sigma-Aldrich

Growth Hormone Releasing Factor human

≥95% (HPLC), powder, synthetic

Synonym(s):

GRF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C215H358N72O66S
CAS Number:
Molecular Weight:
5039.65
MDL number:
UNSPSC Code:
12352200

biological source

synthetic

assay

≥95% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

InChI key

JAHCMOSSKRAPEL-IBFVROBCSA-N

Gene Information

human ... GHRH(2691)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Tyr-Ala-Asp-Ala-Ile-Phe-Thr-Asn-Ser-Tyr-Arg-Lys-Val-Leu-Gly-Gln-Leu-Ser-Ala-Arg-Lys-Leu-Leu-Gln-Asp-Ile-Met-Ser-Arg-Gln-Gln-Gly-Glu-Ser-Asn-Gln-Glu-Arg-Gly-Ala-Arg-Ala-Arg-Leu-NH2

Biochem/physiol Actions

Hypothalamic peptide that stimulates release of somatotropin from anterior pituitary

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service