Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G8642

Sigma-Aldrich

Glutathione S-Transferase from human placenta

lyophilized powder, 25-125 units/mg protein

Synonym(s):

GST, Glutathione S-alkenetransferase, Glutathione S-alkyltransferase, Glutathione S-aralkyltransferase, Glutathione S-aryltransferase, Glutathione S-epoxidetransferase, RX: Glutathione R-transferase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.47

form

lyophilized powder

specific activity

25-125 units/mg protein

composition

Protein, ~40% biuret

UniProt accession no.

storage temp.

−20°C

Gene Information

General description

Glutathione S-transferase (GST) is a major detoxification enzyme, and exists as multiple cytoplasmic and membrane-bound isozymes. Cytoplasmic isoforms of GST are encoded by five genes, namely α, γ, μ, σ and π. α, μ and π are the most abundant forms in mammals. GSTπ is the isoform of this enzyme found in placenta and is composed of 209 amino acids.
Glutathione S-transferase mu 1 (GSTM1) is mapped to human chromosome 1p13.3.

Application

Glutathione S-Transferase from human placenta has been used for the immunization of rabbits for the production of specific anti-π-GST antibodies, to determine whether total anti-π-GST antibodies are more specific and reactive as compared to anti-peptide antibodies.
Glutathione S-Transferase from human placenta has been used in antigen microarray and informatics analysis.

Biochem/physiol Actions

Glutathione S-transferase (GST) catalyzes conjugation reactions during phase II metabolism. This enzyme also confers protection against catabolic products of peroxidized lipid and oxidized DNA, and ROS (reactive oxygen species) or electrophiles.
Glutathione S-transferase (GST) catalyzes the reduction of dehydroascorbic acid to ascorbic acid in the presence of glutathione. GST exhibits neuroprotective action, by eliminating oxidants (exogenous and endogenous), detoxifying therapeutic drugs and other toxins in the environment. Mutation in this gene is known to cause epilepsy. Deletion of the gene may result in cancer.
Glutathione S-transferases are a family of proteins that catalyze the conjugation of reduced glutathione with a variety of hydrophobic chemicals containing electrophilic centers.
Glutathione S-transferases are a protein family catalyzing conjugation of reduced glutathione with various hydrophobic chemicals.

Unit Definition

One unit will conjugate 1.0 μmole of 1-chloro-2,4-dinitrobenzene with reduced glutathione per min at pH 6.5 at 25°C.

Physical form

Lyophilized powder containing Tris buffer salts, reduced glutathione and EDTA.

Analysis Note

Protein determined by biuret.
Purified and assayed by a modification of the method of Simons and Vander Jagt.
Enzymatic activities are based on the conjugation of reduced glutathione with a second substrate. The individual proteins generally have activity with more than one class of substrate.

inhibitor

related product

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glutathione S-transferase M1 and T1 null genotype frequency distribution among four tribal populations of western India
Suthar PC, et al.
Journal of genetics, 1-14 (2018)
Association of GSTT1 and GSTM1 polymorphisms in South Indian Epilepsy Patients
Prabha TS, et al.
Indian Journal of Experimental Biology, 783-787 (2016)
A multi-stage genome-wide association study of bladder cancer identifies multiple susceptibility loci
Rothman N, et al.
Nature Genetics, 42(11), 978-978 (2010)
F Di Modugno et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 53(9-10), 902-910 (1998-11-24)
The main linear epitopes of pi-glutathione transferase (pi-GST, EC 2.5.1.18), an enzyme related to cancer progression in a restricted number of tumours, were identified by testing in ELISA the reactivities of polyclonal anti-pi-GST rabbit sera against a panel of 51
Functional genetic variants of glutathione S-transferase protect against serum ascorbic acid deficiency
Cahill LE, et al.
American Journal of Clinical Nutrition, 90(5), 1411-1417 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service