Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G8535

Sigma-Aldrich

Gastrointestinal Tumor Antigen from human cell culture

≥10,000 units/mL, buffered aqueous solution

Synonym(s):

Cancer-associated, Ca 19-9

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

form

buffered aqueous solution

concentration

≥10,000 units/mL

storage temp.

−20°C

Quality

Highly purified

Unit Definition

One unit is an arbitrary amount related to a reference antigen preparation using CIS-RIA method.

Physical form

Solution in 0.15 M phosphate buffered saline, pH 7.4, containing 0.1% sodium azide

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service