Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G8421

Sigma-Aldrich

GSK3α, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

156-212 nmol/min·mg

mol wt

~81 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... GSK3A(2931)

Biochem/physiol Actions

GSK3a is a multifunctional protein serine kinase, homologous to Drosophila ‘shaggy′ (zeste-white3) and implicated in the control of several regulatory proteins including glycogen synthase and transcription factors (e.g., JUN).GSK3a also plays a role in the WNT and PI3K signaling pathways. Alzheimer disease is associated with increased production and aggregation of amyloid-beta-40 and -42 peptides into plaques. GSK3a is required for maximal production of the beta-amyloid-40 and -42 peptides generated from the amyloid precursor protein by presenilin-dependent gamma-secretase cleavage. In vitro, lithium, a GSK3a inhibitor, blocks the production of the beta-amyloid peptides by interfering with the gamma-secretase step.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glycogen synthase kinase-3: properties, functions, and regulation.
A Ali et al.
Chemical reviews, 101(8), 2527-2540 (2001-12-26)
Christopher J Phiel et al.
Nature, 423(6938), 435-439 (2003-05-23)
Alzheimer's disease is associated with increased production and aggregation of amyloid-beta (Abeta) peptides. Abeta peptides are derived from the amyloid precursor protein (APP) by sequential proteolysis, catalysed by the aspartyl protease BACE, followed by presenilin-dependent gamma-secretase cleavage. Presenilin interacts with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service