Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G7909

Sigma-Aldrich

GROβ human

recombinant, expressed in E. coli, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

biological source

human

recombinant

expressed in E. coli

assay

≥97% (SDS-PAGE)

form

lyophilized powder

potency

0.1-1.0 ng/mL ED50/EC50

mol wt

8 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... CXCL2(2920)

Biochem/physiol Actions

A potent chemoattractant for human neutrophils, but not human monocytes. The biological activity of recombinant, human GROβ?is measured by its ability to induce myeloperoxidase release from cytochalasin B treated neutrophils.

Physical form

Lyophilized from a 0.2 μm filtered solution in 30% acetonitrile and 0.1% trifluoroacetic acid containing 0.5 mg bovine serum albumin

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service