G7903
Glycophorin Predominantly glycophorin A from blood type MM
lyophilized powder
About This Item
biological source
human blood (type MN)
form
lyophilized powder
concentration
20-40% (Lowry)
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... GYPA(2993)
General description
Application
Biochem/physiol Actions
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service