G7657
Anti-GABA Transporter GAT-2 antibody produced in rabbit
affinity isolated antibody, lyophilized powder
About This Item
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
lyophilized powder
contains
0.02% sodium azide
1% bovine serum albumin
species reactivity
rat
technique(s)
immunohistochemistry: suitable
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
rat ... Slc6a13(171163)
Immunogen
Application
Biochem/physiol Actions
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service