Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G6298

Sigma-Aldrich

(RS)-PPG

≥98% (HPLC)

Synonym(s):

(R,S)-4-phosphonophenylglycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10NO5P
CAS Number:
Molecular Weight:
231.14
UNSPSC Code:
12352202
NACRES:
NA.77

assay

≥98% (HPLC)

form

solid

storage condition

desiccated

solubility

DMSO: 2 mg/mL

storage temp.

2-8°C

Biochem/physiol Actions

(RS)-PPG is a potent selective group III mGluR agonist; anticonvulsant and neuroprotectant in vivo.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service