G5758
Glucose-6-Phosphatase from rabbit liver
≥0.05 units/mg protein, Protein ~30 % (balance mostly sucrose)
Synonym(s):
D-Glucose-6-Phosphate Phosphohydrolase
About This Item
Recommended Products
form
powder with sucrose
specific activity
≥0.05 units/mg protein
composition
Protein, ~30% (balance mostly sucrose)
storage temp.
−20°C
General description
Application
Biochem/physiol Actions
Other Notes
a) glucose 6-phosphate + H2O → glucose + Pi
b) PPi + H2O → 2 Pi
c) glucose + PPi or nucleoside-DP or -TP → glucose 6-phosphate + Pi or nucleoside-MP.
Quality
Unit Definition
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
To measure glucose-6-phosphatase activity, the Taussky-Shorr method is used. This method is a spectrophotometric stop-rate determination assay that is measured at 660 nm.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service