Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G5650

Sigma-Aldrich

Grb2 SH2-domain Protein A fusion protein

≥90% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204

recombinant

expressed in E. coli

assay

≥90% (SDS-PAGE)

form

lyophilized powder

composition

Protein, 3-5%

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... GRB2(2885)

Biochem/physiol Actions

A key protein in signal transduction, Grb2 transfers the signal from tyrosine kinases such as epidermal growth factor (EGF) receptor to the small G protein Ras. GRB2 contains an SH2 (Src homology 2) domain which enables binding to tyrosine-phosphorylated receptors. The fusion protein enables detection and purification of tyrosine-phosphorylated proteins (i.e., tyrosine-phosphorylated EGF receptor), utilizing the affinity of protein A for the Fc domain of IgG.

Packaging

Package size based on protein content

Other Notes

A 37 kDa fusion protein containing the SH2 domain of mouse Grb2 (amino acids 50-161) fused to protein A.

Physical form

Contains potassium phosphate and sodium chloride

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Margolis
Progress in biophysics and molecular biology, 62(3), 223-244 (1994-01-01)
The cloning of SH2 domain proteins based on their binding to growth factor receptors is a powerful technique to elucidate new signaling pathways. In some cases the function of these proteins has been quickly ascertained while in others the answers
J P Olivier et al.
Cell, 73(1), 179-191 (1993-04-09)
A Drosophila gene (drk) encodes a widely expressed protein with a single SH2 domain and two flanking SH3 domains, which is homologous to the Sem-5 protein of C. elegans and mammalian GRB2. Genetic analysis suggests that drk function is essential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service