G-proteins are membrane associated heterotrimeric proteins that are comprised of α-, β-, and γ-subunits. These proteins play a key role in signal transduction pathways that regulate cell′s response to several hormones, neuromodulators and ligands. The αz subunit is myristoylated near the N-terminus.
Immunogen
synthetic peptide corresponding to the C-terminal region of Gαx/z.
Application
Anti-Gαx/z antibody produced in rabbit is suitable for western blot analysis at a working dilution of 1:100-1:200 using plasma membrane fractions.
Biochem/physiol Actions
The gene GNAZ (guanine nucleotide binding protein (G protein), αz polypeptide) encodes an α subunit of guanine nucleotide binding proteins. The α-subunit contains a guanine-binding domain that is in its inactive state when it is occupied by GDP. Upon activation, GDP is replaced with GTP, causing the dissociation of the α-subunit from the βγ-subunit complex. This enables the Gα-GTP complex to bind to and regulate specific signaling pathways. GTP is then hydrolyzed, allowing for re-association of the α-subunit with the βγ-subunit complex. When coupled to phospholipase C, the α subunits may have a role in pertussis toxin-insensitive pathways.
The Biochemical journal, 291 ( Pt 2), 349-353 (1993-04-15)
We have examined the post-translational processing of G alpha subunits expressed endogenously in rat PC12 and NG108-15 rat/mouse hybrid cells, and after transfection of cDNA expression constructs into COS cells. Thioester-linked palmitoylation of alpha o, alpha s, alpha q/alpha 11
DNA research : an international journal for rapid publication of reports on genes and genomes, 7(2), 111-120 (2000-05-20)
Heterotrimeric guanine nucleotide binding proteins (G proteins) transduce extracellular signals received by transmembrane receptors to effector proteins. Each subunit of the G protein complex is encoded by a member of one of three corresponding gene families. Currently, 16 different members
The many faces of G protein signaling.
H E Hamm
The Journal of biological chemistry, 273(2), 669-672 (1998-02-14)
Proceedings of the National Academy of Sciences of the United States of America, 90(8), 3675-3679 (1993-04-15)
A small number of membrane-associated proteins are reversibly and covalently modified with palmitic acid. Palmitoylation of G-protein alpha and beta subunits was assessed by metabolic labeling of subunits expressed in simian COS cells or insect Sf9 cells. The fatty acid
In multicellular organisms from Caenorhabditis elegans to Homo sapiens, the maintenance of homeostasis is dependent on the continual flow and processing of information through a complex network of cells. Moreover, in order for the organism to respond to an ever-changing
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.