G5025
β-Galactosidase-biotin labeled from Escherichia coli
lyophilized powder, 350-1200 units/mg protein
About This Item
biological source
Escherichia coli
form
lyophilized powder
specific activity
350-1200 units/mg protein
composition
Protein, ~75% E1%/280
extent of labeling
2-5 mol d-biotin per mol protein
Storage temp.
−20°C
Application
Biochem/physiol Actions
Packaging
Unit Definition
Physical form
Preparation Note
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service