Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G5015

Sigma-Aldrich

Globulins Cohn fraction IV-4 from rat

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202

biological source

rat

form

powder

UniProt accession no.

storage temp.

2-8°C

Gene Information

Other Notes

Predominantly α-globulins

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service