Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G4664

Millipore

George Kimmig selective supplement

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200

application(s)

environmental
food and beverages

microbiology

shipped in

wet ice

storage temp.

2-8°C

Application

An antimicrobial supplement recommended for selective isolation and cultivation of fungi.

Components

Composition: (per vial)
Penicillin G: 20,000 IU
Streptomycin: 20.00 μg

Reconstitution

Sufficient for 500 ml medium

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Jiang et al.
Scientific reports, 4, 7456-7456 (2014-12-17)
SIRT1 could protect degenerative human NP cells against apoptosis, and there were extensive and intimate connection between apoptosis and autophagy. Up to now, the role of autophagy in the process of human IVD degeneration is unclear. We sought to explore

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service