Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G4128

Sigma-Aldrich

Glycine t-butyl ester hydrochloride

Synonym(s):

t-Butyl glycinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO2 · HCl
CAS Number:
Molecular Weight:
167.63
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

−20°C

SMILES string

Cl.CC(C)(C)OC(=O)CN

InChI

1S/C6H13NO2.ClH/c1-6(2,3)9-5(8)4-7;/h4,7H2,1-3H3;1H

InChI key

OSWULUXZFOQIRU-UHFFFAOYSA-N

replaced by

Product No.
Description
Pricing

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ming-Qing Hua et al.
Chemical communications (Cambridge, England), 47(5), 1631-1633 (2010-12-01)
An efficient, catalytic, diastereo- and enantioselective conjugate addition of N-(diphenylmethylene)glycine tert-butyl ester to β-aryl substituted enones was realized in the presence of 1 mol% of newly desired dinuclear N-spiro-ammonium salts, affording functionalized α-amino acid derivatives in 57-98% yields with high
Baptiste Thierry et al.
Molecular diversity, 9(4), 277-290 (2005-11-29)
Cross-linked polystyrene-bound and poly(ethylene glycol)-bound phase-transfer catalysts as well as homopolymers of cinchona alkaloid derivatives have been synthesised. Both soluble and insoluble polymers have been investigated. The enantioselective alkylation of N-diphenyl methylene glycine t-butyl ester has been successfully carried out
S Sarhan et al.
Arzneimittel-Forschung, 34(6), 687-690 (1984-01-01)
The synergistic amplification of the anticonvulsant effects of direct and indirect GABA agonists by glycine has previously been demonstrated. We show in the present report that the anticonvulsant effect of vinyl GABA, a GABA-T (4-aminobutyrate: 2-oxoglutarate aminotransferase) inhibitor with antiepileptic
M Nakoji et al.
Organic letters, 3(21), 3329-3331 (2001-10-12)
[reaction: see text]. A chiral phase-transfer catalyst has been applied to the asymmetric allylation of the tert-butyl glycinate-benzophenone Schiff base with various allylic acetates for the first time to give the allylated products in good yields and with comparable to
V De Filippis et al.
Biochemistry, 37(39), 13507-13515 (1998-09-30)
Hirudin is the most potent and specific inhibitor of thrombin, a key enzyme in the coagulation process existing in equilibrium between its procoagulant (fast) and anticoagulant (slow) form. In a previous study, we described the solid-phase synthesis of a Trp3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service