Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G3919

Sigma-Aldrich

Anti-Groucho3 antibody produced in rabbit

affinity isolated antibody, ammonium sulfate suspension

Synonym(s):

Anti-TLE3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

ammonium sulfate suspension

mol wt

antigen 83.3 kDa

species reactivity

human, rodent

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... TLE3(7090)
mouse ... Tle3(21887)
rat ... Tle3(84424)

Immunogen

a synthetic peptide corresponding to human Groucho3 (TLE3) (amino acids 295-311) conjugated to BSA.

Application

Anti-Groucho3 antibody produced in rabbit is suitable for immunohistochemistry and immunoblotting at a working dilution of 1:200 to 1:1000.

Biochem/physiol Actions

Groucho3 (TLE3) (Transducin-like enhancer protein 3, ESG3) is a member of the Groucho family of transcriptional corepressors and inhibits the transcriptional activation mediated by β-catenin and TCF family members in Wnt signaling.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service