Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G3387

Sigma-Aldrich

Globulins Cohn fraction IV human

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202

biological source

human

assay

≥90% (biuret)

form

powder

impurities

HIV and HBsAg, source material tested negative

storage temp.

2-8°C

Other Notes

Predominantly α- and β-globulins

Preparation Note

Coprecipitation of Cohn Fractions IV-I and IV-4

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

  1. How does the storage temperature relate to shipping conditions?

    The storage conditions that a Sigma-Aldrich catalog and label recommend for products are deliberately conservative. For many products, long-term storage at low temperatures will increase the time during which they are expected to remain in specification and therefore are labeled accordingly. Where short-term storage, shipping time frame, or exposure to conditions other than those recommended for long-term storage will not affect product quality, Sigma-Aldrich will ship at ambient temperature. The products sensitive to short-term exposure to conditions other than their recommended long-term storage are shipped on wet or dry ice. Ambient temperature shipping helps to control shipping costs for our customers. At any time, our customers can request wet- or dry-ice shipment, but the special handling is at customer expense if our product history indicates that the product is stable for regular shipment. See Shipping and Storage for more information.

  2. Which document(s) contains shelf-life or expiration date information for a given product?

    If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.

  3. How do I get lot-specific information or a Certificate of Analysis?

    The lot specific COA document can be found by entering the lot number above under the "Documents" section.

  4. How do I find price and availability?

    There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote.  USA customers:  1-800-325-3010 or view local office numbers.

  5. What is the Department of Transportation shipping information for this product?

    Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product. 

  6. What type of globulins are found in Product No. G3387, Globulins Cohn fraction IV human?

    We report the globulin and albumin content per our testing by agarose electrophoresis. This percentage is reported on the certificate of analysis (CofA) for each lot of this product. The predominant globulins that are present are alpha- and beta-globulins.

  7. What is the protein content of Product No. G3387, Globulins Cohn fraction IV human?

    Our specification for this product requires a protein content by Biuret at ≥ 90%. 

  8. Do you provide the transferrin content for Product No. G3387, Globulins Cohn fraction IV human?

    No, we have not tested this product for its transferrin content.

  9. My question is not addressed here, how can I contact Technical Service for assistance?

    Ask a Scientist here.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service