G3171
Glycinin from Glycine max (soybean)
soybean storage protein
About This Item
Recommended Products
biological source
Glycine max (soybean)
assay
≥80% (HPLC)
form
lyophilized powder
storage temp.
−20°C
Gene Information
soybean ... GY1(547901) , GY2(547900) , GY3(547463) , GY4(100101841) , GY5(547452)
General description
Biochem/physiol Actions
signalword
Danger
hcodes
Hazard Classifications
Resp. Sens. 1 - Skin Sens. 1
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service