Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G2670

Sigma-Aldrich

Anti-G Protein-Coupled Receptor AGR9 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-GPCR AR9

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 3 μg/mL using human adrenal tissue

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OXER1(165140)

General description

AGR9 encodes a 483 amino acid protein with seven stretches of hydrophobic amino acids, which represents a G protein-coupled receptor. It is expressed in the cardiovascular, central nervous and digestive systems.

Immunogen

synthetic peptide corresponding to the C-terminal domain of human G protein-coupled receptor AGR9, conjugated to KLH. The immunizing peptide has 100% identity with the mouse gene.

Application

Anti-G Protein-Coupled Receptor AGR9 antibody produced in rabbit is suitable for immunohistochemistry at a working concentration of 3μg/mL using formalin-fixed, paraffin-embedded sections human adrenal tissue.

Biochem/physiol Actions

AGR9 exhibits rhodopsin-like receptor activity. Dibutyryl cyclic AMP and the β-adrenergic agonist isoproterenol, mimic the effect of forskolin. Forskolin causes a marked and transient decrease in the steady-state level of AGR9 mRNA.

Physical form

Solution in phosphate buffered saline, pH 7.7, containing 0.01% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service