Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G1170

Sigma-Aldrich

Anti-G Protein-Coupled Receptor TG1019 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-GPCR TG1019

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 30 μg/mL using liver

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... OXER1(165140)

General description

G Protein-Coupled Receptor belongs to the largest known receptor family. It is characterized by seven transmembrane domains with an extracellular N terminus and a cytoplasmic C terminus domain. They are further divided into distinct subfamilies depending on their different types of ligands and according to sequence homologies. It is highly expressed in liver, kidney, peripheral leukocyte, lung, and spleen, than in other tissues.

Immunogen

synthetic peptide corresponding to the C-terminus of human G-protein-coupled receptor TG1019, conjugated to KLH.

Application

Anti-G Protein-Coupled Receptor TG1019 antibody is suitable for immunohistochemistry (formalin-fixed, paraffin-embedded sections) at a concentration of 30μg/mL using liver.

Biochem/physiol Actions

G Protein-Coupled Receptor has the ability to alter variety of extracellular signals such as photons, odorants, biogenic amines, peptides, hormone proteins, proteases, nucleotides, and lipids.

Physical form

Solution in phosphate buffered saline, pH 7.7, containing 0.01% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carol E Jones et al.
Molecular pharmacology, 63(3), 471-477 (2003-02-28)
Using a bioinformatics approach, we have isolated a novel G-protein-coupled receptor (GPCR), R527, and have demonstrated that this receptor shows no significant homology to previously deorphanized GPCRs. Quantitative reverse transcription-polymerase chain reaction analysis of the expression of GPCR R527 indicated
Takeshi Hosoi et al.
The Journal of biological chemistry, 277(35), 31459-31465 (2002-06-18)
We have conducted an in silico data base search for and cloned a novel G-protein-coupled receptor (GPCR) named TG1019. Dot and Northern blotting analyses showed that transcripts of the novel GPCR were expressed in various tissues except brain, and the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service