Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G0920

Sigma-Aldrich

Anti-Gonadotropin-Releasing Hormone Receptor antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-GnRHR

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1-2 μg/mL using human pituitary

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GNRHR(2798)
mouse ... Gnrhr(14715)
rat ... Gnrhr(81668)

Immunogen

synthetic peptide corresponding to the third cytoplasmic loop of human gonadotropin-releasing hormone receptor conjugated to KLH. The immunizing peptide has between 90% homology with the rat gene and 95% homology with the mouse gene.

Application

Anti-Gonadotropin-Releasing Hormone Receptor antibody is suitable for immunohistochemistry (formalin-fixed, paraffin-embedded sections) at a concentration of 1-2μg/mL using human pituitary.

Biochem/physiol Actions

GnRHR is a seven transmembrane domain receptor belonging to the rhodopsin-like G protein-coupled receptor superfamily. It lacks a cytoplasmic C-terminal domain, which makes it different from other G-protein coupled receptors. It is expressed on the surface of pituitary gonadotrope cells as well as lymphocytes, breast, ovary, and prostate. It stimulates the action of gonadotropin secretion from the pituitary and thereby controls gametogenesis and steroidogenesis in the gonads.

Physical form

Solution in phosphate buffered saline containing 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kathleen R Sedgley et al.
The Journal of endocrinology, 191(3), 625-636 (2006-12-16)
Gonadotropin-releasing hormone receptors (GnRHRs) are expressed in gonadotropes and several extra-pituitary sites. They are assumed to be cell surface proteins but the human (h) GnRHR lacks features favoring plasma membrane localization and receptor location varies with cell type. When expressed
Chi Keung Cheng et al.
Endocrine reviews, 26(2), 283-306 (2004-11-25)
In human beings, two forms of GnRH, termed GnRH-I and GnRH-II, encoded by separate genes have been identified. Although these hormones share comparable cDNA and genomic structures, their tissue distribution and regulation of gene expression are significantly dissimilar. The actions
L Chi et al.
Molecular and cellular endocrinology, 91(1-2), R1-R6 (1993-02-01)
A cDNA encoding the human GnRH receptor (GnRHR) has been cloned and functionally expressed in both Xenopus oocytes and COS-1 cells. The 2160 bp cDNA encodes a 328 amino acid protein with a predicted amino acid sequence that is 90%

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service