Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G0670

Sigma-Aldrich

Anti-Galanin Receptor 3 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-GalR3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 4 μg/mL using human brain, neurons and glia

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... GALR3(8484)
mouse ... Galr3(14429)
rat ... Galr3(29235)

Immunogen

synthetic peptide corresponding to the C-terminal domain of human galanin receptor 3, conjugated KLH. The immunizing peptide has 100% homology with the rat and mouse gene.

Application

Anti-Galanin Receptor 3 antibody is suitable for immunohistochemistry (formalin-fixed, paraffin-embedded sections) at a concentration of 4μg/mL using human brain, neurons and glia.

Biochem/physiol Actions

Galanin, a small amino acid peptide, is widely expressed in human testis and also in adrenal gland and pancreas to lesser extent. It exerts a broad range of physiological effects. Galanin Receptor 3 is associated with various processes such as hormone release, feeding behavior, smooth muscle contractility and somatosensory neuronal function. Three distinct galanin receptor (GALR) subtypes, designated GALR1, 2, and 3, have been found.

Physical form

Solution in phosphate buffered saline, pH 7.7, containing 0.01% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T P Iismaa et al.
Annals of the New York Academy of Sciences, 863, 56-63 (1999-02-03)
Human galanin receptor subtypes GALR1, GALR2, and GALR3 are encoded by separate genes that are located on human chromosomes 18q23, 17q25.3, and 22q13.1, respectively. The exon:intron organization of the gene encoding GALR2 (GALNR2) and GALR3 (GALNR3) is conserved, with exon
K E Smith et al.
The Journal of biological chemistry, 273(36), 23321-23326 (1998-08-29)
The neuropeptide galanin has been implicated in the regulation of processes such as nociception, cognition, feeding behavior, and hormone secretion. Multiple galanin receptors are predicted to mediate its effects, but only two functionally coupled receptors have been reported. We now
T A Branchek et al.
Trends in pharmacological sciences, 21(3), 109-117 (2000-02-26)
The neuropeptide galanin, which is widely expressed in brain and peripheral tissues, exerts a broad range of physiological effects. Pharmacological studies using peptide analogues have led to speculation about multiple galanin receptor subtypes. Since 1994, a total of three G-protein-coupled
L F Kolakowski et al.
Journal of neurochemistry, 71(6), 2239-2251 (1998-12-01)
Galanin is a 29- or 30-amino acid peptide with wide-ranging effects on hormone release, feeding behavior, smooth muscle contractility, and somatosensory neuronal function. Three distinct galanin receptor (GALR) subtypes, designated GALR1, 2, and 3, have been cloned from the rat.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service