synthetic peptide representing the sequence of the C1 splice variant.
application
Anti-Glutamate Receptor NMDAR1 (NR1) (Splice Variant Insert C1) antibody is suitable for immunohistochemistry (frozen sections) at a dilution of 1:1000-1:2000 and western blot at 1:1000 dilution using rat and mouse tissue.
Biochem/physiol Actions
NMDA receptors (NMDAR) are glutamate-gated ion channels. It is sensitive to N-methyl-D-aspartate (NMDA) and consists of several homologous subunits (NMDAR1). NMDAR is highly expressed in the central nervous system and involved in excitatory synaptic transmission as well as synaptic plasticity. The activity of NMDA receptors are modulated by glycine. It exhibits high calcium permeability and unique voltage-dependent sensitivity to magnesium. It has been reported that phosphorylated NMDA receptors may regulate their function and play a role in many forms of synaptic plasticity such as long-term potentiation.
Physical form
Lyophilized powder from 5 mM ammonium bicarbonate, so some residual salt may be present
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
N-methyl-D-aspartate (NMDA) receptors are highly expressed in the central nervous system and are involved in excitatory synaptic transmission as well as synaptic plasticity. Despite considerable structural and biophysical research, the mechanism behind activation of the NMDA receptor is still poorly
The NMDA (N-methyl D-aspartate) receptors in the brain play a critical role in synaptic plasticity, synaptogenesis and excitotoxicity. Molecular cloning has demonstrated that NMDA receptors consist of several homologous subunits (NMDAR1, 2A-2D). A variety of studies have suggested that protein
Science (New York, N.Y.), 256(5060), 1217-1221 (1992-05-22)
The N-methyl D-aspartate (NMDA) receptor subtype of glutamate-gated ion channels possesses high calcium permeability and unique voltage-dependent sensitivity to magnesium and is modulated by glycine. Molecular cloning identified three complementary DNA species of rat brain, encoding NMDA receptor subunits NMDAR2A
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.