F9762
Anti-Human IgM (μ-chain specific)−FITC antibody produced in goat
affinity isolated antibody, buffered aqueous solution
About This Item
Recommended Products
biological source
goat
Quality Level
conjugate
FITC conjugate
antibody form
affinity isolated antibody
antibody product type
secondary antibodies
clone
polyclonal
form
buffered aqueous solution
storage condition
protect from light
technique(s)
immunofluorescence: suitable (1:32 - 1:64 using Hep2 cells)
storage temp.
−20°C
target post-translational modification
unmodified
Looking for similar products? Visit Product Comparison Guide
General description
Anti-Human IgM (μ-chain specific)-FITC antibody specifically binds to mu-chain of human IgM. After immunospecific purification, anti-human IgM is conjugated to fluorescein isothiocyanate (FITC), in an alkaline reaction. The antibody preparation is specific for human IgM when tested against purified human IgA, IgG, IgM, Bence Jones κ and λ myeloma proteins. The product is also determined to have no cross reactivity with mouse or rat serum proteins.
Specificity
Immunogen
Application
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service