Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F9549

Sigma-Aldrich

Fibroblast Growth Factor Receptor-2α (IIIc)/Fc Chimera human

>90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

Synonym(s):

FGFR-2α (IIIc)/Fc Chimera

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

human

recombinant

expressed in NSO cells

assay

>90% (SDS-PAGE)

form

lyophilized powder

potency

0.637 ng/mL

mol wt

~66 kDa

packaging

pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

<1 EU/μg

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... FGFR2(2263)

General description

FGFR2 (fibroblast growth factor receptor 2) is a member of the tyrosine kinase receptor family called FGFR. This gene is localized to human chromosome 10q26, and is composed of 2 exons.[1] Its ligand-binding domain in present in its extracellular region, which is composed of three immunoglobulin (Ig)-like domains. It spans the membrane once, and its tyrosine kinase domain is localized to its intracellular region. Alternative splicing of this gene gives rise to two isoforms- epithelial (FGFR2IIIb) and mesenchymal (FGFR2IIIc).[2]

Biochem/physiol Actions

FGFR2 (fibroblast growth factor receptor 2) facilitates tumor growth, invasiveness, metastasis and angiogenesis. It plays an essential role in the development of mammary glands, and its expression is elevated in breast cancers. Studies in North Indian population suggest that polymorphisms in this gene might be linked to susceptibility to breast cancer.[1] It is anti-apoptotic in nature, and suppresses p53-mediated apoptosis, by inducing the elevation of MDM2 levels.[3] Mutations in this gene are linked to mild Crouzon syndrome and craniosynostosis.[2]

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline

Analysis Note

The bioactivity is measured by its ability to inhibit human fibroblast growth factor acidic-dependent proliferation of NR6 cells.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aimee L Fenwick et al.
BMC medical genetics, 15, 95-95 (2014-09-02)
Mutations of fibroblast growth factor receptor 2 (FGFR2) account for a higher proportion of genetic cases of craniosynostosis than any other gene, and are associated with a wide spectrum of severity of clinical problems. Many of these mutations are highly
Zahra Saadatian et al.
Asian Pacific journal of cancer prevention : APJCP, 15(18), 7955-7958 (2014-10-09)
Breast cancer is the most common cancer among women in the world. In Iran, the incidence of breast cancer is on the increase. We here studied the association of rs1219648 in FGFR2 and rs1042522 in TP53 and their interaction in
Sarah Siddiqui et al.
PloS one, 9(10), e110426-e110426 (2014-10-22)
Genome-Wide Association Studies (GWAS) have identified Fibroblast growth factor receptor 2 (FGFR2) as a candidate gene for breast cancer with single nucleotide polymorphisms (SNPs) located in intron 2 region as the susceptibility loci strongly associated with the risk. However, replicate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service