F9304
Anti-FBI-1/Pokemon antibody produced in rabbit
IgG fraction of antiserum, buffered aqueous solution
Synonym(s):
Anti-FBI-1, Anti-LRF, Anti-POK Erythroid Myeloid Ontogenic Factor, Anti-ZBTB7, Anti-Zinc Finger- and BTB Domain-containing Protein 7
About This Item
biological source
rabbit
conjugate
unconjugated
antibody form
IgG fraction of antiserum
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen 75 kDa
species reactivity
human
technique(s)
immunoprecipitation (IP): 2-4 μL using A431 cell lysates
western blot (chemiluminescent): 1:4,000-1:6,000 using K-562 lysates
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
Gene Information
human ... ZBTB7A(51341)
Immunogen
Application
Biochem/physiol Actions
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
antibody
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service