F7885
Filtration kit replacement filters
filter size 0.2 μm × 0.2 μm , low volume
Synonym(s):
filtration kit, replacement filter
All Photos(1)
About This Item
UNSPSC Code:
41100000
description
effective filtration area 500 cm2 78 in2
sterility
sterile; γ-irradiated
filter size
0.2 μm × 0.2 μm , low volume
General description
Disposable ready-to-use replacement filter capsules for the Liquid Media Filtration Kit (FK-1) to afford flexibility for your applications. All filter capsule materials have been evaluated for biosafety in accordance with U.S.P. (United States Pharmacopoeia) Class VI Plastics Tests to ensure safety of materials. All filters described are intended for use in positive pressure filtration systems at pressures up to 50 psi. Individual filters or serial combinations of filters can be used to pre-filter and/or sterilize Tissue Culture media (without serum) or reagents in the volumes indicated. Individual filter efficiency and throughput will vary with the product being filtered. Products such as serum, medium containing serum, or contaminated liquids will reduce the capacity of the filter
Two layers of low protein-binding media in a polycarbonate capsule. For sterilization of up to 20L of Tissue Culture media (without serum) or reagents.
Choose from one of the most recent versions:
Certificates of Analysis (COA)