F7776
Monoclonal Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2)−FITC antibody produced in mouse
clone MAPK-YT, purified immunoglobulin, buffered aqueous solution
Synonym(s):
Monoclonal Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2)
About This Item
biological source
mouse
conjugate
FITC conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
MAPK-YT, monoclonal
form
buffered aqueous solution
mol wt
antigen ERK-1 44 kDa
antigen ERK-2 42 kDa
species reactivity
human, Caenorhabditis elegans, Xenopus, Drosophila, hamster, rat, bovine, mouse, yeast
technique(s)
direct immunofluorescence: 1:20 using cultured human foreskin fibroblasts (paraformaldehyde-fixation)
isotype
IgG1
UniProt accession no.
shipped in
dry ice
Storage temp.
−20°C
Gene Information
human ... MAPK1(5594) , MAPK3(5595)
mouse ... Mapk1(26413) , Mapk3(26417)
rat ... Mapk1(116590) , Mapk3(50689)
Specificity
Immunogen
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service