Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F7551

Sigma-Aldrich

Fractalkine, Extracellular Domain from mouse

>97% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder

Synonym(s):

CX3CL-1, Neurotactin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

mouse

recombinant

expressed in baculovirus infected Sf21 cells

assay

>97% (SDS-PAGE)

form

lyophilized powder

potency

~0.06-0.6 μg/mL ED50

mol wt

~90 kDa by SDS-PAGE
calculated mol wt 34 kDa

packaging

pkg of 25 μg

storage condition

avoid repeated freeze/thaw cycles (Do not store in a frost-free freezer.)

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Cx3cl1(20312)

General description

CX3CL1 (chemokine (C-X3-C motif) ligand 1) or fractalkine is a membrane protein, which is the only member of CXC3 family. Its chemokine domain (CKD) anchors it to the cell membrane through its mucin-like stalk. It acts as a ligand for the G-protein coupled receptor CX3CR1, which is found on smooth muscle cells, T-cells, natural killer (NK) cells and monocytes. Fractalkine is expressed on epithelial, endothelial and smooth muscle cells and neurons.[1]

Biochem/physiol Actions

CX3CL1 (chemokine (C-X3-C motif) ligand 1) or fractalkine, along with CX3CR1 is involved in various disorders such as atherosclerosis, multiple sclerosis, neuropathic pain and rheumatic disorders. It facilitates the proliferation and survival of primary human vascular smooth muscle cells in an epidermal growth factor receptor (EGR)–dependent manner. It plays an essential role in cell survival and promotes the survival of cells such as monocytes, T-cells and microglia. In humans, it is thought to be involved in atherogenesis, and thus, might have potential as a target in treatment of cardiovascular disease.[1] In patients with sickle cell disease (SCD), the serum levels of this chemokine is increased, thus implicating it in the pathogenesis of inflammation associated with SCD.[2]
Leukocyte chemoattractant protein and member of the chemokine superfamily possessing a CX3C motif.
Recombinant extracellular fractalkine simulates the shed form of the chemokine and chemoattracts neutrophils and T cells.
Simulates the shed form of the chemokine and chemoattracts neutrophils and T cells.

Other Notes

Extracellular domain (amino acids 1-337) fused to a 6X histidine tag at the C-terminus.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 1.25 mg of bovine serum albumin.

Analysis Note

The biological activity is measured by its ability to chemoattract freshly isolated peripheral blood lymphocytes.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selma Unal et al.
International journal of hematology, 101(2), 114-118 (2014-12-07)
In the present study, we examined the role of fractalkine (Fkn), a member of the chemokine family, in the pathogenesis of sickle cell disease (SCD). Eighty-seven children with sickle cell disease and 55 healthy children were enrolled in the study.
Gemma E White et al.
Arteriosclerosis, thrombosis, and vascular biology, 34(12), 2554-2562 (2014-11-02)
The CX3C chemokine fractalkine (CX3CL1) has a critical role in the development of atherogenesis because apolipoprotein-E-deficient mice lacking CX3CL1 or its receptor CX3CR1 develop smaller plaques and polymorphisms in CX3CR1 are associated with altered risk of cardiovascular disease. CX3CR1 is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service