Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

F7297

Sigma-Aldrich

H-Trisaccharide

synthetic

Synonym(s):

α-Fuc-(1→2)-β-Gal-(1→4)-GlcNAc, 2′-Fucosyl-N-acetyllactosamine, Blood group H trisaccharide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H35NO15
Molecular Weight:
529.49
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

assay

≥95%

form

solid

storage temp.

2-8°C

SMILES string

C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@@H](O)[C@@H](CO)O[C@H]2O[C@H]3[C@H](O)[C@@H](NC(C)=O)C(O)O[C@@H]3CO)[C@@H](O)[C@H](O)[C@@H]1O

InChI

1S/C20H35NO15/c1-5-10(25)13(28)15(30)19(32-5)36-17-14(29)11(26)7(3-22)34-20(17)35-16-8(4-23)33-18(31)9(12(16)27)21-6(2)24/h5,7-20,22-23,25-31H,3-4H2,1-2H3,(H,21,24)/t5-,7+,8+,9+,10+,11-,12+,13+,14-,15-,16+,17+,18?,19-,20-/m0/s1

InChI key

PHTAQVMXYWFMHF-GJGMMKECSA-N

Application

Blood group H trisaccharide may be used to isolate, identify and characterize blood group trisaccharide specific lectins and ligands such as Cymbosema roseum lectin II (CRL II) and Ralstonia solanacearum lectin (RSL).

Other Notes

Human blood group H type trisaccharide.
To gain a comprehensive understanding of our extensive range of Oligosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bernard C Collins et al.
Structure (London, England : 1993), 25(11), 1667-1678 (2017-10-11)
High-quality reagents to study and detect glycans with high specificity for research and clinical applications are severely lacking. Here, we structurally and functionally characterize several variable lymphocyte receptor (VLR)-based antibodies from lampreys immunized with O erythrocytes that specifically recognize the
Lifeng Liu et al.
Plant physiology, 168(2), 549-560 (2015-04-15)
The predominant structure of the hemicellulose xyloglucan (XyG) found in the cell walls of dicots is a fucogalactoXyG with an XXXG core motif, whereas in the Poaceae (grasses and cereals), the structure of XyG is less xylosylated (XXGGn core motif)

Articles

O-Linked glycoproteins are usually large proteins with a molecular mass of >200 kDa. Glycosylation generally occurs in high-density clusters and may represent as much as 50-80% of the overall mass.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service