Key Documents
F7147
Cryopreservation supplies
forceps
About This Item
UNSPSC Code:
41103000
NACRES:
NB.22
material
stainless steel
L
6 in.
General description
Curved, grooved end allows easier manipulation of cryopreservation straws, vials and other small objects. Avoids directly handling cryopreserved cells or embryos, thereby minimizing premature warming.
Choose from one of the most recent versions:
Certificates of Analysis (COA)