Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

F6931

Sigma-Aldrich

Anti-FGL1 (281-295) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-FGL1 fibrinogen-like 1, Anti-HFREP1, Anti-HP-041, Anti-LFIRE1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~34 kDa

species reactivity

human

technique(s)

western blot: 1:500-1:2,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FGL1(2267)

Immunogen

synthetic peptide corresponding to amino acids 281-295 of human FGL1

Application

Anti-FGL1 (281-295) antibody is suitable for western blot at a dilution of 1:500-1:2,000.
Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about us and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Biochem/physiol Actions

FGL1 belongs to the fibrinogen family, the principal protein in formation of blood clots in vertebrates. It exists as a multimeric protein structure comprising two copies of three homologous polypeptide chains (α2, β2, γ2). It is reacted with the thrombin-like enzyme, forming a fibrin-like substance in blood coaggulation. The downregulation of the protein is significantly associated with the differentiation of hepatocellular carcinomas (HCC). It has been reported that in deregulation of FGL1, it acts in cell growth and tumorigenicity in vitro and in vivo. Studies conclude that FGL1 may contribute in hepatocarcinogenesis.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Y Park et al.
Archives of andrology, 38(1), 29-36 (1997-01-01)
This investigation was conducted to examine the presence of fibrinogen-like substance and thrombin-like enzyme in human semen (human seminal plasma) after liquefaction. The human seminal plasma contained small amounts of respective substances which are absorbed on anti-fibrinogen and anti-thrombin III
Jun Yan et al.
Oncogene, 23(10), 1939-1949 (2004-02-26)
We have previously identified several novel genes, which are differentially expressed among human normal liver and hepatocellular carcinomas (HCCs). The full-length liver fibrinogen-related gene-1 (LFIRE-1) cDNA was cloned from the human normal liver cDNA library. LFIRE-1 is highly homologous to
H Ichijo et al.
The Journal of biological chemistry, 268(19), 14505-14513 (1993-07-05)
We have previously identified and purified transforming growth factor-beta 1 (TGF-beta 1)-binding proteins from porcine uterus membranes (Ichijo, H., Rönnstrand, L., Miyagawa, K., Ohashi, H., Heldin, C.-H., and Miyazono, K. (1991) J. Biol. Chem. 266, 22459-22464). One of these TGF-beta

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service