Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F5929

Sigma-Aldrich

Anti-FBG4 (FBXO17) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-F Box 4

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~31 kDa by calculation

species reactivity

mouse, human, rat

technique(s)

western blot: 1:500-1:1,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

peptide corresponding to the human FBG4 protein (amino acids 70-85).

Application

Anti-FBG4 (FBXO17) antibody is suitable for western blot at a dilution of 1:500-1:1,000.

Biochem/physiol Actions

F-box protein, is the critical component of conserved ubiquitin–protein ligase complex SCF (Skp1/Cdc53-Cullin1/F-box), and acts as a substrate recognition component of the ubiquitin ligase complex SCF (Skp-Cullin-F-box). It is expressed in human tissues including kidney, liver, brain and muscle tissues. It is associated with various critical biological processes such as in regulating the cell cycle, immune response, signaling cascades and developmental programs. The proteins have a bipartite structure and the core complex, whereas divergent protein-protein interaction motifs selectively bind their cognate substrates.

Physical form

Solution of 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gennady P Ilyin et al.
Gene, 296(1-2), 11-20 (2002-10-18)
F-box proteins, a critical component of the evolutionary conserved ubiquitin-protein ligase complex SCF (Skp1/Cdc53-Cullin1/F-box), recruit substrates for ubiquitination and consequent degradation through their specific protein-protein interaction domains. Here, we report the identification of full-length cDNAs encoding three novel human F-box
Laura Pontano Vaites et al.
Molecular and cellular biology, 31(22), 4513-4523 (2011-09-14)
Skp1-Cul1-F-box (SCF) E3 ubiquitin ligase complexes modulate the accumulation of key cell cycle regulatory proteins. Following the G(1)/S transition, SCF(Fbx4) targets cyclin D1 for proteasomal degradation, a critical event necessary for DNA replication fidelity. Deregulated cyclin D1 drives tumorigenesis, and
Margaret S Ho et al.
Journal of biomedical science, 13(2), 181-191 (2006-02-08)
The eukaryotic protein degradation pathway involves the ubiquitin (Ub) modification of substrates targeted for degradation by the 26S proteasome. The addition of Ub, a process called ubiquitination, is mediated by enzymes including the E3 Ub ligases which transfer the Ub
K L Craig et al.
Progress in biophysics and molecular biology, 72(3), 299-328 (1999-12-03)
The ubiquitin system of intracellular protein degradation controls the abundance of many critical regulatory proteins. Specificity in the ubiquitin system is determined largely at the level of substrate recognition, a step that is mediated by E3 ubiquitin ligases. Analysis of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service